414557

Sigma-Aldrich

Ethyl (pentafluorobenzoyl)acetate

98%

Linear Formula:
C6F5COCH2CO2C2H5
CAS Number:
Molecular Weight:
282.16
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.464 (lit.)

bp

103-104 °C/4 mmHg (lit.)

density

1.43 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1c(F)c(F)c(F)c(F)c1F

Pictograms

FlameExclamation mark

Signal Word

Warning

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3272 3 / PGIII

WGK Germany

3

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.