MilliporeSigma
  • 415170
All Photos(1)

415170

Sigma-Aldrich

Barium yttrium tungsten oxide

99.9%

Linear Formula:
Ba3Y2WO9
CAS Number:
Molecular Weight:
917.63
MDL number:
PubChem Substance ID:

Assay

99.9%

form

powder

particle size

<5 μm

SMILES string

O=[Ba].O=[Ba].O=[Ba].O=[W](=O)=O.O=[Y]O[Y]=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service