MilliporeSigma
All Photos(1)

416088

Sigma-Aldrich

Poly(acrylic acid-co-maleic acid) sodium salt

average Mw ~70,000

CAS Number:
MDL number:

mol wt

average Mw ~70,000

SMILES string

OC(=O)C=C.OC(=O)\C=C\C(=O)O[Na]

InChI

1S/C4H2O3.C3H4O2.Na/c5-3-1-2-4(6)7-3;1-2-3(4)5;/h1-2H;2H,1H2,(H,4,5);/q;;+1/p-1

InChI key

LDLISKWWEPKHAT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

∼2.2:1 mole ratio of AA:MA

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service