MilliporeSigma
All Photos(1)

419745

Sigma-Aldrich

7-Methoxyflavone

99%

Empirical Formula (Hill Notation):
C16H12O3
CAS Number:
Molecular Weight:
252.26
MDL number:
PubChem Substance ID:

assay

99%

mp

110-112 °C (lit.)

SMILES string

COc1ccc2C(=O)C=C(Oc2c1)c3ccccc3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service