419761

Sigma-Aldrich

6-Methoxyflavanone

98%

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
MDL number:
PubChem Substance ID:

assay

98%

mp

141-143 °C (lit.)

SMILES string

COc1ccc2OC(CC(=O)c2c1)c3ccccc3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service