MilliporeSigma
All Photos(1)

430307

Sigma-Aldrich

Dimer acid diglycidyl ester

CAS Number:
MDL number:

eq. wt.

390-490 Epoxide

refractive index

n20/D 1.486 (lit.)

viscosity

400 cP(25 °C)(lit.)

density

0.98 g/mL at 25 °C (lit.)

Application

Epoxy flexibilizer.

Features and Benefits

Improves impact strength and toughness.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service