All Photos(1)

449091

Sigma-Aldrich

(4-Chlorophenylthio)acetic acid

98%

Linear Formula:
ClC6H4SCH2CO2H
CAS Number:
Molecular Weight:
202.66
MDL number:

assay

98%

mp

104-107 °C (lit.)

SMILES string

OC(=O)CSc1ccc(Cl)cc1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service