All Photos(1)

451231

Sigma-Aldrich

Samarium(III) bromide

anhydrous, beads, −10 mesh, 99.99%

Synonym(s):
Samarium tribromide, Tribromosamarium
Linear Formula:
SmBr3
CAS Number:
Molecular Weight:
390.07
EC Number:
MDL number:
PubChem Substance ID:

grade

anhydrous

assay

99.99%

form

beads

particle size

−10 mesh

mp

700 °C (lit.)

SMILES string

Br[Sm](Br)Br

InChI

1S/3BrH.Sm/h3*1H;/q;;;+3/p-3

InChI key

ICKYUJFKBKOPJT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service