464082

Sigma-Aldrich

6-O-(Triisopropylsilyl)-D-glucal

95%

Empirical Formula (Hill Notation):
C15H30O4Si
CAS Number:
Molecular Weight:
302.48
MDL number:
PubChem Substance ID:

assay

95%

optical activity

[α]22/D −2°, c = 1 in chloroform

refractive index

n20/D 1.489 (lit.)

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CC(C)[Si](OC[C@H]1OC=C[C@@H](O)[C@@H]1O)(C(C)C)C(C)C

InChI

1S/C15H30O4Si/c1-10(2)20(11(3)4,12(5)6)19-9-14-15(17)13(16)7-8-18-14/h7-8,10-17H,9H2,1-6H3/t13-,14-,15+/m1/s1

InChI key

VYZHAYQIZWKLNP-KFWWJZLASA-N

Related Categories

Packaging

1 g in glass bottle

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Certificate of Analysis

Certificate of Origin

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service