MilliporeSigma
All Photos(1)

473626

Sigma-Aldrich

4-Bromo-2,2-diphenylbutyric acid

98%

Linear Formula:
BrCH2CH2C(C6H5)2CO2H
CAS Number:
Molecular Weight:
319.19
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

mp

131 °C (dec.) (lit.)

SMILES string

OC(=O)C(CCBr)(c1ccccc1)c2ccccc2

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service