Documents

477346

Sigma-Aldrich

(S)-(−)-1,2-Epithiododecane

Synonym(s):
(S)-(−)-Decylthiirane
Empirical Formula (Hill Notation):
C12H24S
CAS Number:
Molecular Weight:
200.38
MDL number:
PubChem Substance ID:

optical activity

[α]25/D −44°, neat

refractive index

n20/D 1.472 (lit.)

bp

119-120 °C/4 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC[C@H]1CS1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service