MilliporeSigma
All Photos(1)

482528

Sigma-Aldrich

ArgoPore-Wang

70-120 mesh, extent of labeling: 0.5-1.0 mmol/g OH loading, highly cross-linked

Synonym(s):
4-Benzyloxybenzyl alcohol, polymer-bound, p-Alkoxybenzyl alcohol, polymer-bound, Wang resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene
MDL number:

crosslinking

highly cross-linked

extent of labeling

0.5-1.0 mmol/g OH loading

particle size

70-120 mesh

Looking for similar products? Visit Product Comparison Guide

Legal Information

ArgoPore is a trademark of Argonaut Technologies, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service