All Photos(1)

482854

Sigma-Aldrich

(S,S)-(−)-1-(1-Naphthyl)-2-(2-naphthyl)-1,2-ethanediol

98%

Linear Formula:
C10H7CH(OH)CH(OH)C10H7
CAS Number:
Molecular Weight:
314.38
MDL number:
PubChem Substance ID:

assay

98%

optical activity

[α]24/D −147°, c = 1 in chloroform

optical purity

ee: 98% (HPLC)

mp

100-105 °C (lit.)

SMILES string

O[C@H]([C@@H](O)c1cccc2ccccc12)c3ccc4ccccc4c3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service