All Photos(1)

494046

Sigma-Aldrich

Cadmium hydroxide

98%

Linear Formula:
Cd(OH)2
CAS Number:
Molecular Weight:
146.43
EC Number:
MDL number:

assay

98%

mp

232 °C (lit.)

density

4.79 g/mL at 25 °C (lit.)

SMILES string

O[Cd]O

InChI

1S/Cd.2H2O/h;2*1H2/q+2;;/p-2

InChI key

PLLZRTNVEXYBNA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service