All Photos(1)

496308

Sigma-Aldrich

[2,3,3,3-Tetrafluoro-2-(trifluoromethoxy)propyl]oxirane

97%

Synonym(s):
[2,3,3,3-Tetrafluoro-2-(trifluoromethoxy)propyl]oxirane
Empirical Formula (Hill Notation):
C6H5F7O2
CAS Number:
Molecular Weight:
242.09
MDL number:
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.316 (lit.)

bp

108 °C (lit.)

density

1.496 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)OC(F)(CC1CO1)C(F)(F)F

InChI

1S/C6H5F7O2/c7-4(5(8,9)10,1-3-2-14-3)15-6(11,12)13/h3H,1-2H2

InChI key

YLTWQTPPDBTMJI-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service