Documents

496324

Sigma-Aldrich

(2,2,3,3,4,4,4-Heptafluorobutyl)oxirane

96%

Empirical Formula (Hill Notation):
C6H5F7O
CAS Number:
Molecular Weight:
226.09
MDL number:

assay

96%

refractive index

n20/D 1.318 (lit.)

bp

111 °C (lit.)

density

1.462 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)CC1CO1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service