All Photos(1)

498564

Sigma-Aldrich

2-Ethylphenylzinc iodide solution

0.5 M in THF

Linear Formula:
C2H5C6H4ZnI
CAS Number:
Molecular Weight:
297.45
MDL number:
PubChem Substance ID:

concentration

0.5 M in THF

density

0.978 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCc1ccccc1[Zn]I

InChI

1S/C8H9.HI.Zn/c1-2-8-6-4-3-5-7-8;;/h3-6H,2H2,1H3;1H;/q;;+1/p-1

InChI key

RDLZIUOCWMAAGW-UHFFFAOYSA-M

Related Categories

Packaging

50 mL in Sure/Seal™

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service