All Photos(1)

513768

Sigma-Aldrich

Bromide, polymer-supported

macroporous, 40-60 mesh, extent of labeling: 3.5 mmol/g loading

Synonym(s):
Amberlyst® A-26 Br--form, Ambersep® 900 bromide form, Bromide on polymer support
Linear Formula:
C6H4CH2N(CH3)3Br
PubChem Substance ID:

extent of labeling

3.5 mmol/g loading

particle size

40-60 mesh

SMILES string

[Br-].C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)Cc3ccccc3

Application

Polymer-supported reagent used in the preparation of alkyl bromides from alkyl chlorides and in the chlorobromination of oleofins and acetylenes.

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Ambersep is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Certificate of Analysis

Certificate of Origin

Bongini, A et al.
Synthesis, 143-143 (1980)
Cainelli, G. et al.
Synthesis, 472-472 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service