Documents

523445

Sigma-Aldrich

Cobalt

wire, diam. 0.1 mm, 99.995%

Synonym(s):
Cobalt element, Cobalt-59
Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
EC Number:
MDL number:

assay

99.995%

form

wire

resistivity

6.24 μΩ-cm, 20°C

diam.

0.1 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service