MilliporeSigma
All Photos(1)

525812

Sigma-Aldrich

Bis(4-chlorophenoxy)acetic acid

96%

Linear Formula:
(ClC6H4O)2CHCO2H
CAS Number:
Molecular Weight:
313.13
EC Number:
MDL number:
PubChem Substance ID:

assay

96%

mp

138-140 °C (lit.)

SMILES string

OC(=O)C(Oc1ccc(Cl)cc1)Oc2ccc(Cl)cc2

InChI

1S/C14H10Cl2O4/c15-9-1-5-11(6-2-9)19-14(13(17)18)20-12-7-3-10(16)4-8-12/h1-8,14H,(H,17,18)

InChI key

ZKNSZZXBPSICFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

A N Kondrat'ev et al.
Anesteziologiia i reanimatologiia, (2)(2), 27-30 (2008-06-11)
The paper provides the results of a complex experimental and clinical study of the effects of infusion solutions of hydroxyethyl starch (HES 200/0.5--Refortan and HES 130/0.4--Voluven) on hemostatic and systemic hemodynamic parameters in patients operated on for neurosurgical pathology of
V V Moskalev et al.
Anesteziologiia i reanimatologiia, (1)(1), 32-35 (2005-04-21)
The paper comparatively evaluates the efficiency of the use of physiological sodium chloride solution, freshly-frozen plasma, 6% and 10% solutions of hydroxyethyl starch to compensate for circulating blood volume deficit in 80 neonatal infants aged 1-3 days who had hypovolemia
A N Kondrat'ev et al.
Vestnik khirurgii imeni I. I. Grekova, 166(6), 56-60 (2008-04-17)
The investigation has shown that solutions of hydroxyethyl starch included in complex infusion therapy facilitate stabilization of hemodynamics in neurosurgical patients during surgery. Refortan exerts a more pronounced effect on the indices of hemodynamics as compared with voluven, but its
Michael Winterhalter et al.
European journal of medical research, 15(9), 383-389 (2010-10-19)
the constantly growing amount of different kinds of colloid fluids necessitates comparative investigations with regards to the safety and effectivity in clinical use of these preparations. Hence we compared three colloid fluids in an observational study. The objective was the
V O Kryliuk
Klinichna khirurhiia, (2)(2), 67-70 (2013-05-28)
The peculiarities of course of endogenous intoxication (EI) in severe combined abdominal trauma were studied up in experiment. There was established, that in the laboratory animals, to whom the blood loss compensation was done, using colloid-hyperosmolar solution, the El severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service