Documents

527033

Sigma-Aldrich

(4-Chlorophenyl)diphenylsulfonium triflate

Synonym(s):
(4-Chlorophenyl)diphenylsulfonium trifluoromethanesulfonate
Empirical Formula (Hill Notation):
C19H14ClF3O3S2
CAS Number:
Molecular Weight:
446.89
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

mp

108-112 °C (lit.)

λmax

239 nm

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Clc1ccc(cc1)[S+](c2ccccc2)c3ccccc3

Application

Cationic photoinitiator. Photoacid generator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26-36

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.