All Photos(1)

529079

Sigma-Aldrich

4-Chloro-2-fluorocinnamic acid, predominantly trans

96%

Linear Formula:
ClC6H3(F)CH=CHCO2H
CAS Number:
Molecular Weight:
200.59
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

96%

mp

213-216 °C (lit.)

SMILES string

OC(=O)\C=C\c1ccc(Cl)cc1F

InChI

1S/C9H6ClFO2/c10-7-3-1-6(8(11)5-7)2-4-9(12)13/h1-5H,(H,12,13)/b4-2+

InChI key

FVLPOWWRHAOKMT-DUXPYHPUSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service