All Photos(1)

533858

Sigma-Aldrich

1,2-Dibromo-1,1,2-trichloroethane

98%

Linear Formula:
Cl2C(Br)CH(Cl)Br
CAS Number:
Molecular Weight:
291.20
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.569 (lit.)

bp

94-95 °C/10 mmHg (lit.)

density

1.701 g/mL at 25 °C (lit.)

SMILES string

ClC(Br)C(Cl)(Cl)Br

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service