All Photos(1)

537942

Sigma-Aldrich

3-(Thiocyanato)propyl-functionalized silica gel

200-400 mesh, extent of labeling: 0.8 mmol/g loading

CAS Number:
MDL number:

extent of labeling

0.8 mmol/g loading

particle size

200-400 mesh
60 Å

surface area

500 m2/g

SMILES string

O=[SiH2].CCCSC#N

Application

Scavenges amines.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service