All Photos(1)

547549

Sigma-Aldrich

StratoSpheres PL-PPZ (Piperazine) resin

50-100 mesh, extent of labeling: >5.0 mmol/g loading, 1 % cross-linked

Synonym(s):
Piperazine, polymer-bound
MDL number:
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

>5.0 mmol/g loading

particle size

50-100 mesh

Looking for similar products? Visit Product Comparison Guide

Legal Information

StratoSpheres is a trademark of Agilent Technologies, Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service