555991

Sigma-Aldrich

3,4,5-Trifluorobenzenesulfonyl chloride

97%

Linear Formula:
F3C6H2SO2Cl
CAS Number:
Molecular Weight:
230.59
MDL number:
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.4970 (lit.)

bp

76-77 °C (lit.)

density

1.658 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(cc(F)c1F)S(Cl)(=O)=O

InChI

1S/C6H2ClF3O2S/c7-13(11,12)3-1-4(8)6(10)5(9)2-3/h1-2H

InChI key

HSEHKULCGSMHAE-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Supp Hazards

Reacts violently with water.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2924 8(3) / PGIII

WGK Germany

3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C

Certificate of Analysis

Certificate of Origin

Articles
Aryl sulfonyl chloride derivatives are frequently used in parallel synthesis to synthesize sulfonamides and sulfonate linkages.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.