All Photos(1)

557005

Sigma-Aldrich

1,2-O-Cyclohexylidene-α-D-glucofuranose

95%

Empirical Formula (Hill Notation):
C12H20O6
CAS Number:
Molecular Weight:
260.28
MDL number:
PubChem Substance ID:

assay

95%

mp

163-168 °C (lit.)

SMILES string

[H][C@@]1(O[C@@H]2OC3(CCCCC3)O[C@@H]2[C@H]1O)[C@H](O)CO

InChI

1S/C12H20O6/c13-6-7(14)9-8(15)10-11(16-9)18-12(17-10)4-2-1-3-5-12/h7-11,13-15H,1-6H2/t7-,8+,9-,10-,11-/m1/s1

InChI key

KAUCUKASFLXURN-RCZSTQMZSA-N

Related Categories

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service