All Photos(1)

562947

Sigma-Aldrich

2-Amino-N-ethyl-N-phenylbenzenesulfonamide

97%

Synonym(s):
2-Aminobenzene-N-ethyl-N-phenyl sulfonamide
Linear Formula:
NH2C6H4SO2N(CH2CH3)C6H5
CAS Number:
Molecular Weight:
276.35
EC Number:
MDL number:
PubChem Substance ID:

assay

97%

mp

56-60 °C (lit.)

SMILES string

CCN(c1ccccc1)S(=O)(=O)c2ccccc2N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service