All Photos(1)

597023

Sigma-Aldrich

(S)-(+)-1,1′-Bi-2-naphthyl diacetate

96%

Empirical Formula (Hill Notation):
C24H18O4
CAS Number:
Molecular Weight:
370.40
MDL number:
PubChem Substance ID:

assay

96%

optical activity

[α]20/D +16°, c = 1% in chloroform

optical purity

ee: 97% (HPLC)

mp

107-111 °C (lit.)

SMILES string

CC(=O)Oc1ccc2ccccc2c1-c3c(OC(C)=O)ccc4ccccc34

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service