634670

Sigma-Aldrich

Strontium titanate

single crystal substrate, <110>, ≥99.99% trace metals basis

Synonym(s):
Strontium titanium trioxide
Linear Formula:
SrTiO3
CAS Number:
Molecular Weight:
183.49
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

description

single side polished

assay

≥99.99% trace metals basis

dielectric constant

~300

hardness

6 (, Mohs)

L × W × thickness

10 mm × 10 mm × 0.5 mm

mp

2060 °C (lit.)

solubility

H2O: insoluble

density

4.81 g/mL at 25 °C (lit.)

semiconductor properties

<110>

SMILES string

[Sr++].[O-][Ti]([O-])=O

InChI

1S/3O.Sr.Ti/q;2*-1;+2;

InChI key

VEALVRVVWBQVSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

loss tangent at 10 GHz = ~5 × 10-4 (300 K), ~3 × 10-4 (77 K)
Single crystal strontium titanate substrate lattice has a good lattice match with most perovskite structured materials. It is an excellent substrate from the epitaxial growth of many oxide this films and high-temperature superconductors. It also has applications in optical windows and as a high-quality sputtering target.

Physical form

cubic (a = 3.905 Å)

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Chemically driven nanoscopic magnetic phase separation at the SrTiO(3) (001)/La(1-x) Sr(x) CoO(3) interface.
Maria A Torija et al.
Advanced materials (Deerfield Beach, Fla.), 23(24), 2711-2715 (2011-04-21)
L Avilés Félix et al.
Nanotechnology, 23(49), 495715-495715 (2012-11-17)
The transport properties of ultra-thin SrTiO(3) (STO) layers grown over YBa(2)Cu(3)O(7) electrodes were studied by conductive atomic force microscopy at the nano-scale. A very good control of the barrier thickness was achieved during the deposition process. A phenomenological approach was...
Archana Pandey et al.
ACS nano, 7(1), 117-125 (2012-12-04)
Novel PMMA-STO-CNT matrices were created by opened-tip vertically aligned multiwalled carbon nanotubes (VA-MWCNTs) with conformal coatings of strontium titanate (STO) and poly(methyl methacrylate) (PMMA). Emission threshold of 0.8 V/μm was demonstrated, about 5-fold lower than that of the as-grown VA-MWCNTs....
Troy K Townsend et al.
ACS nano, 6(8), 7420-7426 (2012-07-24)
SrTiO(3) (STO) is a large band gap (3.2 eV) semiconductor that catalyzes the overall water splitting reaction under UV light irradiation in the presence of a NiO cocatalyst. As we show here, the reactivity persists in nanoscale particles of the...
Ralf Moos et al.
Sensors (Basel, Switzerland), 11(4), 3439-3465 (2011-12-14)
Resistive oxygen sensors are an inexpensive alternative to the classical potentiometric zirconia oxygen sensor, especially for use in harsh environments and at temperatures of several hundred °C or even higher. This device-oriented paper gives a historical overview on the development...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.