MilliporeSigma
All Photos(1)

69200

Sigma-Aldrich

4-Methylstyrene

≥99.0%, contains ~0.005% 4-tert-butylcatechol as inhibitor

Synonym(s):
4-Vinyltoluene
Linear Formula:
CH3C6H4CH=CH2
CAS Number:
Molecular Weight:
118.18
Beilstein:
1209317
EC Number:
MDL number:
PubChem Substance ID:

vapor pressure

<1 mmHg ( 20 °C)

assay

≥99.0%

form

liquid

autoignition temp.

959 °F

contains

~0.005% 4-tert-butylcatechol as inhibitor

expl. lim.

5.3 %

refractive index

n20/D 1.542 (lit.)
n20/D 1.542

bp

170-175 °C (lit.)

density

0.895 g/mL at 20 °C
0.897 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(C=C)cc1

InChI

1S/C9H10/c1-3-9-6-4-8(2)5-7-9/h3-7H,1H2,2H3

InChI key

JLBJTVDPSNHSKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service