MilliporeSigma
All Photos(1)

701505

Sigma-Aldrich

Chloro(triphenylphosphite)gold

97%

Empirical Formula (Hill Notation):
C18H15AuClO3P
Molecular Weight:
542.70
MDL number:
PubChem Substance ID:

assay

97%

reaction suitability

reagent type: catalyst
core: gold

mp

97-102 °C

SMILES string

Cl[Au].O(P(Oc1ccccc1)Oc2ccccc2)c3ccccc3

InChI

1S/C18H15O3P.Au.ClH/c1-4-10-16(11-5-1)19-22(20-17-12-6-2-7-13-17)21-18-14-8-3-9-15-18;;/h1-15H;;1H/q;+1;/p-1

InChI key

DADNGSZZNVQSMJ-UHFFFAOYSA-M

Application

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service