MilliporeSigma
All Photos(1)

704881

Sigma-Aldrich

Chloro(dimethylphenylphosphine)gold

97%

Synonym(s):
(Dimethylphenylphosphine)gold chloride
Empirical Formula (Hill Notation):
C8H11AuClP
CAS Number:
Molecular Weight:
370.57
MDL number:
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

98-102 °C

SMILES string

Cl[Au].CP(C)c1ccccc1

InChI

1S/C8H11P.Au.ClH/c1-9(2)8-6-4-3-5-7-8;;/h3-7H,1-2H3;;1H/q;+1;/p-1

InChI key

JUNOUWMLMVXRDI-UHFFFAOYSA-M

Packaging

250 mg in glass bottle
1 g in glass bottle

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service