705853

Sigma-Aldrich

Tetradecahydro-4,7-phenanthroline

greener alternative

95%

Empirical Formula (Hill Notation):
C12H22N2
CAS Number:
Molecular Weight:
194.32
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

95%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

density

1.044 g/mL at 25 °C

SMILES string

C1CNC2CCC3NCCCC3C2C1

InChI

1S/C12H22N2/c1-3-9-10-4-2-8-14-12(10)6-5-11(9)13-7-1/h9-14H,1-8H2

InChI key

SAJIWHKJMCWNFA-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Packaging

500 mg in glass insert

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi

Risk Statement

37/38-41-52/53

Safety Statement

26-39

RIDADR

NONH for all modes of transport

WGK Germany

3

Flash Point(F)

>230 °F

Flash Point(C)

>110 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.