712361

Sigma-Aldrich

(2S,2′S)-1,1′-Bis(2-pyridinylmethyl)-2,2′-bipyrrolidine tetrahydrochloride

95%

Synonym(s):
(S,S)-BPBP tetrahydrochloride, (S,S′)-PDP tetrahydrochloride, 2,2′-[(2S,2′S)-[2,2′-Bipyrrolidine]-1,1′-diylbis(methylene)]bis-pyridine tetrahydrochloride
Empirical Formula (Hill Notation):
C20H26N4 · 4HCl
Molecular Weight:
468.29
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥94.5% (HPLC)
94.5-105.5% (AT)
95%

optical purity

enantiomeric excess: ≥98.0%

SMILES string

Cl.Cl.Cl.Cl.C1C[C@@H]([C@@H]2CCCN2Cc3ccccn3)N(C1)Cc4ccccn4

InChI

1S/C20H26N4.4ClH/c1-3-11-21-17(7-1)15-23-13-5-9-19(23)20-10-6-14-24(20)16-18-8-2-4-12-22-18;;;;/h1-4,7-8,11-12,19-20H,5-6,9-10,13-16H2;4*1H/t19-,20-;;;;/m0..../s1

Inchi Key

WTQUEUGYNCZETF-NRMJSFJRSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.