715549

Sigma-Aldrich

N,N′-Bis[(11bR)-3,5-dihydro-3,5-dimethyl-4-oxido-4H-dinaphtho[2,1-d:1′,2′-f][1,3,2]diazaphosphepin-4-yl]-N,N′-dimethyl-1,5-pentanediamine

95%

Empirical Formula (Hill Notation):
C51H52N6O2P2
CAS Number:
Molecular Weight:
842.95
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

95%

optical activity

[α]22/D −320°, c = 0.5 in methanol

impurities

≤20 wt. % benzene

mp

139-146 °C

storage temp.

2-8°C

SMILES string

CN(CCCCCN(C)P1(=O)N(C)c2ccc3ccccc3c2-c4c(ccc5ccccc45)N1C)P6(=O)N(C)c7ccc8ccccc8c7-c9c(ccc%10ccccc9%10)N6C

InChI

1S/C51H52N6O2P2/c1-52(60(58)54(3)44-30-26-36-18-8-12-22-40(36)48(44)49-41-23-13-9-19-37(41)27-31-45(49)55(60)4)34-16-7-17-35-53(2)61(59)56(5)46-32-28-38-20-10-14-24-42(38)50(46)51-43-25-15-11-21-39(43)29-33-47(51)57(61)6/h8-15,18-33H,7,16-17,34-35H2,1-6H3

InChI key

HTIRTTKKZTYOST-UHFFFAOYSA-N

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.