MilliporeSigma
All Photos(1)

717290

Sigma-Aldrich

Chloro(methyldiphenylphosphine)gold(I)

95%

Empirical Formula (Hill Notation):
C13H13AuClP
CAS Number:
Molecular Weight:
432.64
MDL number:
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

116-118 °C

SMILES string

Cl[Au].CP(c1ccccc1)c2ccccc2

InChI

1S/C13H13P.Au.ClH/c1-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13;;/h2-11H,1H3;;1H/q;+1;/p-1

InChI key

CAOYFWWUVVBZOV-UHFFFAOYSA-M

Application

Reactant for synthesis of:
  • Bis(alkynethiolate)gold(I) derivatives
  • Cinylgold(I) phosphane complexes

Reactant for:
  • Oxidative addition of iodine to gold(I) complexes
  • Formation of three coordinate gold(I) complexes

Packaging

250 mg in glass bottle

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service