717754

Sigma-Aldrich

1,1′-Bis((2R,5R)-2,5-diisopropylphospholano)ferrocene(cyclooctadiene)rhodium(I) tetrafluoroborate

97%

Linear Formula:
C38H60FeP2Rh ⋅ BF4
CAS Number:
Molecular Weight:
824.39
MDL number:
PubChem Substance ID:

assay

97%

optical activity

[α]20/D 332°, c = 0.1 in methylene chloride

mp

235-238 °C

SMILES string

[Fe].[Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.CC(C)[C@H]2CC[C@H](C(C)C)P2[C]3[CH][CH][CH][CH]3.CC(C)[C@H]4CC[C@H](C(C)C)P4[C]5[CH][CH][CH][CH]5

InChI

1S/2C15H24P.C8H12.BF4.Fe.Rh/c2*1-11(2)14-9-10-15(12(3)4)16(14)13-7-5-6-8-13;1-2-4-6-8-7-5-3-1;2-1(3,4)5;;/h2*5-8,11-12,14-15H,9-10H2,1-4H3;1-2,7-8H,3-6H2;;;/q;;;-1;;+1/b;;2-1-,8-7-;;;/t2*14-,15-;;;;/m11..../s1

InChI key

CEECLGREXPXHOQ-VXMPMTHESA-N

Related Categories

Packaging

100 mg in clear glass bottle

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service