722111

Sigma-Aldrich

5-Oxazolecarboxylic acid

97%

Synonym(s):
Oxazole-5-carboxylic acid
Empirical Formula (Hill Notation):
C4H3NO3
CAS Number:
Molecular Weight:
113.07
MDL number:
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

97%

mp

226-230 °C

SMILES string

OC(=O)c1cnco1

InChI

1S/C4H3NO3/c6-4(7)3-1-5-2-8-3/h1-2H,(H,6,7)

InChI key

QCGMEWVZBGQOFN-UHFFFAOYSA-N

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Sodium Tanshinone II-A Sulfonate (DS-201) Induces Vasorelaxation of Rat Mesenteric Arteries via Inhibition of L-Type Ca
Xiao-Dong Zhang et al.
Frontiers in pharmacology, 9, 62-62 (2018-02-20)
Máté Katona et al.
American journal of physiology. Gastrointestinal and liver physiology, 310(3), G193-G204 (2015-11-27)
We have previously shown that chenodeoxycholic acid (CDCA) strongly inhibits pancreatic ductal HCO3 (-) secretion through the destruction of mitochondrial function, which may have significance in the pathomechanism of acute pancreatitis (AP). Ursodeoxycholic acid (UDCA) is known to protect the...
Na Wang et al.
European journal of pharmacology, 870, 172873-172873 (2019-12-24)
Hydroxy-safflor yellow A (HSYA) can exert a variety of effects upon the vascular system. However, the underlying mechanisms are not clear. The present study is to investigate its vasodilating effect and the mechanisms. Wistar-Kyoto (WKY) rats and spontaneously hypertensive rats...
Yui Takashina et al.
Nutrients, 10(10) (2018-09-23)
A chronic magnesium deficiency may be one of the causes of lifestyle-related diseases such as hypertension and diabetes. Serum Mg2+ concentration is strictly controlled by the reabsorption pathway in the renal tubules, but little is known about how Mg2+ reabsorption...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.