MilliporeSigma
All Photos(1)

724076

Sigma-Aldrich

2,3-Dichloropyridine-4-carboxylic acid

97%

Synonym(s):
2,3-Dichloro-4-pyridinecarboxylic acid, 2,3-Dichloroisonicotinic acid
Empirical Formula (Hill Notation):
C6H3Cl2NO2
CAS Number:
Molecular Weight:
192.00
MDL number:
PubChem Substance ID:

assay

97%

form

solid

mp

203-207 °C

SMILES string

OC(=O)c1ccnc(Cl)c1Cl

InChI

1S/C6H3Cl2NO2/c7-4-3(6(10)11)1-2-9-5(4)8/h1-2H,(H,10,11)

InChI key

VGKZZKKTZKRCPA-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service