735957

Sigma-Aldrich

Ytterbium(III) chloride hexahydrate

99.9999% trace metals basis

Synonym(s):
Ytterbium trichloride hydrate, Ytterbium(III) chloride·6hydrate
Linear Formula:
YbCl3 · 6H2O
CAS Number:
Molecular Weight:
387.49
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99.9999% trace metals basis

mp

180 °C (dec.) (lit.)

density

2.58 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Yb](Cl)Cl

InChI

1S/3ClH.6H2O.Yb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

LEYFXTUKPKKWMP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Uliana Kostiv et al.
Beilstein journal of nanotechnology, 6, 2290-2299 (2016-01-07)
NaYF4:Yb(3+)/Er(3+) nanoparticles were synthesized by thermal decomposition of lanthanide trifluoroacetates using oleylamine (OM) as both solvent and surface binding ligand. The effect of reaction temperature and time on the properties of the particles was investigated. The nanoparticles were characterized by...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.