741051

Sigma-Aldrich

Zinc(II) oxo acetate

95%

Synonym(s):
Basic Zinc Acetate, Tetrameric Zn cluster, Zinc, hexakisμ-(acetato-κO,O′)-μ4-oxotetrazinc
Linear Formula:
C12H18Zn4O13
CAS Number:
Pricing and availability is not currently available.

assay

95%

mp

257.2 °C

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Steffen Hausdorf et al.
The journal of physical chemistry. A, 112(33), 7567-7576 (2008-07-26)
The contributions of terephthalic acid and Zn(2+)-coordinated water in N,N-diethylformamide (DEF) to the overall proton activity in the synthesis of MOF-5 (Zn4O(BDC)3, BDC = 1,4-benzenedicarboxylate) were quantitatively determined by combined electrochemical and UV-vis spectroscopic measurements. Structural transformations of zinc carboxylate-based...
Steffen Hausdorf et al.
Journal of the American Chemical Society, 132(32), 10978-10981 (2010-08-12)
A simple two-component procedure was developed to synthesize not only classical zinc-based IRMOFs represented by MOF-5 but also the cobalt and beryllium homologues of this most prominent MOF. The procedure is the first manifestation of mirroring the IRMOF series with...
Steffen Hausdorf et al.
Dalton transactions (Cambridge, England : 2003), 7(7), 1107-1113 (2009-03-27)
An ethynylene diisophthalic acid linker molecule was synthesized and used to form a zinc carboxylate-based metal organic framework (MOF) with very large pores and unit cell volume resulting from the unusual combination of structurally different inorganic units forming the secondary...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.