Documents

87287

Sigma-Aldrich

Tetrahexadecylammonium tetrafluoroborate

≥98.0% (N)

Linear Formula:
[CH3(CH2)15]4N(BF4)
Molecular Weight:
1002.54
MDL number:
PubChem Substance ID:

assay

≥98.0% (N)

impurities

≤0.05% halides (as bromide)

mp

103-107 °C

SMILES string

F[B-](F)(F)F.CCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCC)(CCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCC

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service