94591

Sigma-Aldrich

(S)-Fmoc-3-(trifluoromethyl)-β-Homophe-OH

≥97.0% (HPLC)

Synonym(s):
Fmoc-3-(trifluoromethyl)-L-β-homophenylalanine, (S)-3-(Fmoc-amino)-4-[3-(trifluoromethyl)phenyl]butyric acid
Empirical Formula (Hill Notation):
C26H22F3NO4
CAS Number:
Molecular Weight:
469.45
MDL number:
PubChem Substance ID:

assay

≥97.0% (HPLC)

application(s)

peptide synthesis: suitable

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)C[C@H](Cc1cccc(c1)C(F)(F)F)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C26H22F3NO4/c27-26(28,29)17-7-5-6-16(12-17)13-18(14-24(31)32)30-25(33)34-15-23-21-10-3-1-8-19(21)20-9-2-4-11-22(20)23/h1-12,18,23H,13-15H2,(H,30,33)(H,31,32)/t18-/m0/s1

InChI key

SHSVTUCIAQWRSI-SFHVURJKSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service