All Photos(1)

A17404

Sigma-Aldrich

O-Acetylmandelic chloride

≥95%

Synonym(s):
2-Acetoxy-2-phenylacetyl chloride
Linear Formula:
CH3CO2CH(C6H5)COCl
CAS Number:
Molecular Weight:
212.63
EC Number:
MDL number:
PubChem Substance ID:

assay

≥95%

refractive index

n20/D 1.514 (lit.)

bp

125-130 °C/10 mmHg (lit.)

density

0.943 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OC(C(Cl)=O)c1ccccc1

InChI

1S/C10H9ClO3/c1-7(12)14-9(10(11)13)8-5-3-2-4-6-8/h2-6,9H,1H3

InChI key

BERNQQVIUAZUHY-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service