All Photos(1)

A2203

Sigma-Aldrich

3-Acetamidobenzotrifluoride

97%

Synonym(s):
α,α,α-Trifluoro-m-acetotoluidide
Linear Formula:
CH3CONHC6H4CF3
CAS Number:
Molecular Weight:
203.16
EC Number:
MDL number:

assay

97%

mp

103-106 °C (lit.)

SMILES string

CC(=O)Nc1cccc(c1)C(F)(F)F

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service