MilliporeSigma
All Photos(1)

C3308

Sigma-Aldrich

2-Carbethoxy-5,7-dihydroxy-4′-methoxyisoflavone

Synonym(s):
Ethyl 5,7-dihydroxy-3-(4-methoxyphenyl)-4-oxo-4H-benzo[b]pyran-2-carboxylate
Empirical Formula (Hill Notation):
C19H16O7
CAS Number:
Molecular Weight:
356.33
EC Number:
MDL number:
PubChem Substance ID:

mp

190-193 °C (lit.)

SMILES string

CCOC(=O)C1=C(C(=O)c2c(O)cc(O)cc2O1)c3ccc(OC)cc3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service