All Photos(1)

D86108

Sigma-Aldrich

2-(Diethylamino)acetonitrile

95%

Linear Formula:
(C2H5)2NCH2CN
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:

vapor pressure

760 mmHg ( 170 °C)

assay

95%

impurities

<3% water

refractive index

n20/D 1.426 (lit.)

bp

170 °C (lit.)

density

0.866 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC#N

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service