All Photos(1)

E5409

Sigma-Aldrich

2-Ethoxy-3-methoxybenzaldehyde

98%

Linear Formula:
C2H5OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.536 (lit.)

bp

140 °C/15 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOc1c(OC)cccc1C=O

InChI

1S/C10H12O3/c1-3-13-10-8(7-11)5-4-6-9(10)12-2/h4-7H,3H2,1-2H3

InChI key

DMPUNCUVRGJYGL-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service