All Photos(1)

P1004

Sigma-Aldrich

DL-Penicillamine acetone adduct hydrochloride monohydrate

98%

Synonym(s):
2,2,5,5-Tetramethyl-4-thiazolidinecarboxylic acid hydrochloride, N,S-Isopropylidene-DL-penicillamine 2,2,5,5-Tetramethylthiazolidine-4-carboxylic acid
Empirical Formula (Hill Notation):
C8H15NO2S · HCl · H2O
CAS Number:
Molecular Weight:
243.75
EC Number:
MDL number:

assay

98%

mp

200 °C (dec.) (lit.)

SMILES string

O.Cl.CC1(C)NC(C(O)=O)C(C)(C)S1

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service